Order Entry
Czech Republic
ContactUsLinkComponent
Anti-KRT7 Rabbit Recombinant Antibody [clone: OV-TL 12/30]
Anti-KRT7 Rabbit Recombinant Antibody [clone: OV-TL 12/30]
Catalog # ABNORAB03400.200UG
Supplier:  Abnova
Anti-KRT7 Rabbit Recombinant Antibody [clone: OV-TL 12/30]
Catalog # ABNORAB03400.200UG
Supplier:  Abnova

Specifications

  • Antigen symbol:
    KRT7
  • Clonality:
    Recombinant
  • Clone:
    OV-TL 12/30
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human KRT7.
  • Pk:
    200 µG

Specifications

About this item