Anti-Notch 2 Rabbit Recombinant Antibody [clone: B9]
Catalog # ABNORAB03727.200UG
Supplier: Abnova
Specifications
- Antigen symbol:Notch 2
- Clonality:Recombinant
- Clone:B9
- Host:Rabbit
- Reactivity:Mouse
- Immunogen:Recombinant rabbit monoclonal antibody raised against NRR of mouse Notch 2.
- Pk:200 µG