Anti-TNFRSF9 Rabbit Recombinant Antibody [clone: TKS-1]
ABNORAB03651.200UG
: Abnova
- Antigen symbol:TNFRSF9
- Clonality:Recombinant
- Clone:TKS-1
- Host:Rabbit
- Reactivity:Mouse
- Immunogen:Recombinant rabbit monoclonal antibody raised against mouse Tnfsf9.
- Pk:200 µG