Anti-Isotype control Rabbit Recombinant Antibody [clone: KLH-G9]
Catalog # ABNORAB02803.200UL
Supplier: Abnova
Specifications
- Antigen symbol:Isotype control
- Clonality:Recombinant
- Clone:KLH-G9
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human Isotype control.
- Pk:200 µl