Anti-TAX1BP1 Rabbit Recombinant Antibody [clone: R07-6F6]
ABNORAB01857.100UL
: Abnova
- Antigen symbol:TAX1BP1
- Clonality:Recombinant
- Clone:R07-6F6
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Immunogen:Recombinant rabbit monoclonal antibody raised against human TAX1BP1.
- Pk:100 µl