Anti-ATK1 (pSer473) Rabbit Monoclonal Antibody [clone: B9] (FITC)
ABNOMAB23390.100
: Abnova
- Antigen symbol:ATK1
- Clonality:Monoclonal
- Clone:B9
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- Reactivity:Human
- Modification:pSer473
- Immunogen:Rabbit monoclonal antibody raised against synthetic phosphopeptide of human ATK1.
- Pk:100 AU