Zadání objednávky
Czech Republic
ContactUsLinkComponent
VERISHIELD™ Earmuffs 100 IS
VERISHIELD™ Earmuffs 100 IS
Catalog # DALL1035101-VS
Supplier:  HONEYWELL SAFETY
VERISHIELD™ Earmuffs 100 IS
Catalog # DALL1035101-VS
Supplier:  HONEYWELL SAFETY
Specifications
  • Výška police:
    25
  • Popis:
    VS140 VeriSheild™ earmuff without air flow control, low level/flat, over the head
  • Bal.:
    10
Specifications
About this item
VeriSheild™ is ideal for wearing with other PPE which may be required in particular environments. They offer a choice of styles and attenuation (SNR) levels makes it easy to find the suitable earmuffs for an appropriate level of protection, and comfort.
  • Easy to fit
  • Suitable for wearers with sensitive ear canals
  • Suitable for wearing with hearing aids
  • It helps to keep ears warm
Air flow control and sound management technology that improves employee safety by blocking harmful noise while allowing voice and signals heard more naturally.