Orders LinkContactUsLinkComponent
Safety spectacles, uvex skyguard NT
Safety spectacles, uvex skyguard NT
Supplier:   UVEX
Safety spectacles, uvex skyguard NT
Supplier:  UVEX
Certificates
Safety spectacles, uvex skyguard NT
Safety spectacles, uvex skyguard NT
2 Options Available Below
Product Details & Documents
The dual lens safety glasses were designed to provide an all-round protection. Based on the model Skylite spectacle a conceptually simple, injection moulded TPU frame was added.
  • TPU frame increases wearer protection levels, through minimising the risk of particle ingress
  • Flexible, closed-pore soft components of the frame enable an individual fit
  • Easy to clean
  • Adjustable length of side arms
EN 166-168, EN 170
Certifications: EN 166-168, EN 170
Documents
Product Family Options
Product Information
  • Type
    Colour
    Lenses
    Availability
    Price
  • Anti-UV, anti-scratch, anti-fog
    Blue/grey
    Clear PC
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 450,00 Kč
    450,00 Kč
    1 * 1 KS
       450,00 Kč
  • Anti-UV, anti-scratch
    Blue/grey
    Grey PC
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 356,00 Kč
    356,00 Kč
    1 * 1 KS
       356,00 Kč