Zadání objednávky
Czech Republic
ContactUsLinkComponent
Experimentation kits, blood-based cancer diagnostics
Experimentation kits, blood-based cancer diagnostics
Catalog # EDVO141
Supplier:  EDVOTEK
undefined
Experimentation kits, blood-based cancer diagnostics
Catalog # EDVO141
Supplier:  EDVOTEK
Specifications
  • Popis:
    Blood-based Cancer Diagnostics
  • Bal.:
    1
Specifications
About this item
Cancer cells differ from normal cells by the combinations of proteins that are present on their surfaces. Antibodies against these proteins will specifically bind to cancer cells and not to normal cells. This allows early detection of cancer and potentially a way of delivering cancer therapies. In this simulation experiment the reaction of cancer cell markers and their corresponding antigens are demonstrated.
  • For 10 lab groups
  • Complete in 35 minutes
Delivery: Kit includes instructions, microtitre plates, cancer cell markers, normal cell markers, transfer pipettes and buffer.