ContactUsLinkComponent
DNA Staining System, FlashBlue™, Edvotek
DNA Staining System, FlashBlue™, Edvotek
Catalog # EDVO609
Supplier:  EDVOTEK
CAS Number:  
DNA Staining System, FlashBlue™, Edvotek
Catalog # EDVO609
Supplier:  EDVOTEK
CAS Number:  
Specifications
  • Pk:
    60 mL
  • Description:
    FlashBlue™ DNA Staining System
Specifications
About this item
FlashBlue™ is a DNA stain that offers simple and rapid staining of agarose gels.
  • 10X Concentrate for 1,2 L
Concentrated liquid that, when diluted, can be used for both rapid and overnight staining of DNA fragments.