Order Entry
Czech Republic
ContactUsLinkComponent
Laboratory Brushes
Laboratory Brushes
Catalog # SIALZ281344-1PAK
Supplier:  Merck
Laboratory Brushes
Catalog # SIALZ281344-1PAK
Supplier:  Merck

Specifications

  • Brush head Ø:
    35 mm
  • Total length:
    305 mm
  • Brush head length:
    89 mm
  • Pk:
    4

Specifications

About this item

Brush materials resistant to acids, aromatic and aliphatic hydrocarbons, ketones, ethyl acetate esters, trichloroethylene, and hot water.

  • FDA and USDA approved materials