SteriWare Multi Grip Tweezers
These SteriWare multi grip tweezers operate like conventional tweezers, they have an additional grip feature enabling the easy and secure handling of small vials and containers.
- Made from food grade PS
- Individually bagged to prevent cross contamination
- 100% manufactured and packed in a cleanroom
- Choice of sterile and non sterile
Frequently Bought Together