Hemin chloride (from porcine) ≥96.0% (by HPLC), BioXtra, Sigma-Aldrich®
: Merck
:16009-13-5
:
:C₃₄H₃₂ClFeN₄O₄
:MFCD00010726
:651.95 g/mol
:M-Clarity™ quality level = MQ200
:Refrigerator
:240-140-1
Frequently Bought Together
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
α-Ketoglutaric acid monopotassium salt 97%
: Thermo Fisher Scientific