Order Entry
Czech Republic
ContactUsLinkComponent
Metallised Solvent Resistant Matte Grey Polyester Labels, Property Tag Header, for M6 M7 Printers
Metallised Solvent Resistant Matte Grey Polyester Labels, Property Tag Header, for M6 M7 Printers
  BRDY173863
 :  Brady
undefined
Metallised Solvent Resistant Matte Grey Polyester Labels, Property Tag Header, for M6 M7 Printers
  BRDY173863
 :  Brady

 

  • Type:
    B-428
  • Colour:
    Black on grey
  • Dimensions:
    41,2W×19,05H mm
  • Material:
    Polyester
  • Model:
    M6-35-428-PROP
  • Packed:
    250/roll
  • Pk:
    1 Roll

 

 

Metallised polyester (B-428) labels are designed for applications that utilise barcodes, alphanumerics, graphic symbols and logos and require nameplate-like quality.

  • B-428 is a metallised polyester with a permanent acrylic adhesive and is thermal transfer printable
  • Matte, light grey finish gives labels a rating and serial plate appearance
  • Labels are designed to withstand numerous solvents and variable temperatures when applied to various surfaces
  • UL approved; CSA accepted
  • Shape: Rectangle
  • Service temperature range: −40 to −155,5 °C

Brady materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability.

Application: Asset tracking, circuit board labeling, component and equipment labeling, data and telecommunications labeling, general identification, inventory and inspection labeling.

Resistance properties: Chemical-resistant, corrosion-resistant, humidity-resistant, UV-resistant, weather-resistant.