ContactUsLinkComponent
Aggressive Adhesive Multi-Purpose Polyester Labels with Ribbon for M510, BMP51, BMP53 and M511 Printers
Aggressive Adhesive Multi-Purpose Polyester Labels with Ribbon for M510, BMP51, BMP53 and M511 Printers
  BRDY170859
 :  Brady
undefined
Aggressive Adhesive Multi-Purpose Polyester Labels with Ribbon for M510, BMP51, BMP53 and M511 Printers
  BRDY170859
 :  Brady
  • Colour:
    Black on white
  • Dimensions:
    38,10W mm × 7,62L m
  • Material:
    Polyester
  • Model:
    M5C-1500-422
  • Pk:
    1
  • Adhesive:
    Permanent acrylic
Easily label components with the thermal transfer, glossy permanent polyester label material (B-422). Print almost anywhere with a durable Brady handheld label printer.
  • Versatile - Designed for a wide range of applications such as labeling equipment, circuit boards, panel components and barcodes
  • Tough - Constructed to withstand abrasions, extreme temperatures and solvents such as alcohol, oil and water
  • Adhesive - Made with a permanent acrylic adhesive that sticks to rough, smooth and powder-coated flat surfaces
Ideal for asset tracking, barcode labelling, component and equipment labelling, data and telecommunications labelling, general identification, inventory and inspection labelling applications.
Gloss finish, chemical-resistant, corrosion-resistant, humidity-resistant, UV-resistant, weather-resistant, outdoor use B-422 labels are compatible with Brady printer models M510, BMP51, BMP53, and M511.
 : UL Recognized to UL969, CSA to C22.2 No.0.15-95, RoHS Compliant to 2011/65/EU (Pass 2015/863).