Module for Remote Signalling
Supplier: asecos
Certificates
About this item
The module offers an alarm forwarding via the mobile phone network (SMS/call) and is therefore especially suitable for companies and facilities without a central building management system. In an emergency, the immediate alerting of one or several defined persons is triggered.
- The module is easily installable on-site via plug
- Five programmable telephone numbers
- Configurable message texts
- Collective alarm in case of an emergency or power failure
A SIM card is required for the initial operation of the module, which must be provided by the customer. Alternatively, the included SIM card (for european and UK use only) can be activated via an online portal. A good mobile phone connection is necessary to ensure that the signal can be transmitted.
The remote signalling module is suited for all ION-LINE models.
Frequently Bought Together