ContactUsLinkComponent
Micro-Touch® NitraFree™ Examination Gloves, Nitrile, Powder-Free
Micro-Touch® NitraFree™ Examination Gloves, Nitrile, Powder-Free
Certificates
undefined
Micro-Touch® NitraFree™ Examination Gloves, Nitrile, Powder-Free
Specifications
  • Catalog No:
  • ANSM6034514
  • ANSM6034513
  • ANSM6034512
  • ANSM6034511
  • ANSM6034510
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Material:
  • Nitrile
  • Nitrile
  • Nitrile
  • Nitrile
  • Nitrile
  • Finish:
  • Textured fingertips
  • Textured fingertips
  • Textured fingertips
  • Textured fingertips
  • Textured fingertips
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Finger thickness:
  • 0,12 mm
  • 0,12 mm
  • 0,12 mm
  • 0,12 mm
  • 0,12 mm
  • Palm thickness:
  • 0,08 mm
  • 0,08 mm
  • 0,08 mm
  • 0,08 mm
  • 0,08 mm
  • FDA regulated:
  • +
  • +
  • +
  • +
  • +
  • Sterility:
  • Non sterile
  • Non sterile
  • Non sterile
  • Non sterile
  • Non sterile
  • ASTM No.:
  • D6319
  • D6319
  • D6319
  • D6319
  • D6319