Order Entry
Czech Republic
ContactUsLinkComponent
SAN 400E Dividable Storage Box
SAN 400E Dividable Storage Box
Catalog # MULC50206BAC400ET
Supplier:  MULTIROIR
undefined
SAN 400E Dividable Storage Box
Catalog # MULC50206BAC400ET
Supplier:  MULTIROIR

Specifications

  • Capacity:
    2 L
  • Colour:
    Transparent
  • Material:
    SAN
  • W×D×H:
    External: 400×91×81 mm
    Internal: 365×80×70 mm
  • Pk:
    1
  • Weight:
    250 g

Specifications

About this item

This dividable storage box is a very robust and practical product, as well as being durable and economical.

  • Hygienic: One-piece construction for easy cleaning and decontamination
  • Stackable: Optimises storage space on lab benches or in laboratory cabinets
  • Chemical resistance: Made from SAN, offering excellent resistance to chemicals commonly used in the laboratory
  • Perfect transparency: Allows contents to be identified quickly without handling, reducing the risk of contamination
  • Extended temperature range: Withstands variations from −20 to +80 °C; compatible with laboratory refrigerators and incubators

The SAN 400E dividable storage box is specially designed to meet the stringent requirements of laboratory environments. Its total transparency enables immediate identification of samples and reagents, optimising the efficiency of analytical procedures and reducing the risk of errors. Designed to fit neatly into laboratory workspaces, the bin can be placed safely in storage cabinets, on benches, or shelves, thanks to its balanced and stable shape. The recessed handle makes it easy to handle, even when wearing laboratory gloves, ensuring safe transport of sensitive samples or chemicals.