Biohazard Bags, Autoclavable, Sterile, Double Pouched
KACC3748CBHNPST10D
- Packed:Double pouched, 10 bags/pack
- Colour:Clear
- Seq:03
- Pk:100
- W×L:940×1220 mm
- W×L (imp.):37×48"
Keystone Cleanroom Products Pre-sterilised biohazard bags are useful to decontaminate waste or potentially hazardous materials that are exiting the aseptic production area through the autoclave. The use of sterile biohazard bags eliminates the introduction of contamination into the aseptic production area.
- Ready to use, packed in easy to open Chevron-Peel DuPont™ Tyvek® pouches
- Maximum sterilisation temperature: 135 °C (275 °F)
- Each package is labeled with an expiration date and lot number for complete traceability
Eliminate a common source of contamination.
: Certificates of Conformance sent with each shipment.