Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
AmberChrom™ Na+ form
AmberChrom™ Na+ form 50WX4 200-400 mesh, Supelco®
AmberChrom™ Na+ form 50WX4 200-400 mesh, Supelco®
AmberChrom™ Na+ form 50WX4 200-400 mesh, Supelco®
SIAL44473-100G
:
Merck
:
AmberChrom™ Na+ form 50WX4 200-400 mesh, Supelco®
SIAL44473-100G
:
Merck
:
Pk:
100 g
Seq:
3401
:
:
69011-22-9