Order Entry
Czech Republic
ContactUsLinkComponent
Calcium hydroxide ≥96.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Calcium hydroxide ≥96.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Calcium hydroxide ≥96.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (acidimetric):
    min. 96.0%
  • Substances insolubale in hydrochloric acid:
    max. 0.03%
  • Carbonate (as CaCO3):
    max. 3.0%
  • Chloride (Cl):
    max. 0.005%
  • Sulfur compounds (as SO4):
    max. 0.1%
  • Heavy metals (as Pb):
    max. 0.003%
  • Fe (Iron):
    max. 0.05%
  • K (Potassium):
    max. 0.05%
  • Mg (Magnesium):
    max. 0.5%
  • Na (Sodium):
    max. 0.05%
  • Sr (Strontium):
    max. 0.05%
  • Conform to ACS, Reag. Ph.Eur.:
    pass

Specification Test Results

Related Information

Frequently Bought Together