
Oxalic acid concentrate for dilution to 0.05 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
Catalog # 92510.600
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Pk:210 mL
- Seq:2545
Specifications
Specification Test Results
- Concentration (after dilution to 1000 ml):c(C2H2O4) = 0.05 mol/l ± 0.2% (20°C)
Specification Test Results
About this item
:
:126.07 g/mol
Storage Temperature:Ambient
UN:3265
ADR:8,III
:GHS05
CAS Number:6153-56-6
Formula:HO₂CCO₂H·2H₂O
MDL Number:MFCD00149102