ContactUsLinkComponent
HIGH STAT PLUS ESD Jackets
undefined
HIGH STAT PLUS ESD Jackets
  • Catalog No:
  • ALSO1017456-XL
  • ALSO1017456-2XL
  • ALSO1017456-XS
  • ALSO1017456-S
  • ALSO1017456-M
  • ALSO1017456-L
  • Garment application:
  • Cleanroom, Industrial
  • Cleanroom, Industrial
  • Cleanroom, Industrial
  • Cleanroom, Industrial
  • Cleanroom, Industrial
  • Cleanroom, Industrial
  • Risk area:
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • ESD
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Fabric:
  • 68% polyester/30,5% cotton/1,5% carbon
  • 68% polyester/30,5% cotton/1,5% carbon
  • 68% polyester/30,5% cotton/1,5% carbon
  • 68% polyester/30,5% cotton/1,5% carbon
  • 68% polyester/30,5% cotton/1,5% carbon
  • 68% polyester/30,5% cotton/1,5% carbon
  • Weight:
  • 175 g/m²
  • 175 g/m²
  • 175 g/m²
  • 175 g/m²
  • 175 g/m²
  • 175 g/m²
Frequently Bought Together