DermiFlex Ultra Gloves
Supplier: Portwest
Certificates
About this item
The DermiFlex Ultra provides additional protection against water, oil, and grease due to the additional ³/₄ dip compared to the A350.
- Flexible sandy nitrile coating offers great grip in wet and dry conditions
- ³/₄ dipped for increased protection
- 15 gauge liner for extra dexterity
- OEKO-TEX® STANDARD 100 approved
- Lightweight and comfortable
Low linting construction for minimal contamination, with a breathable seamless liner. The super light 15 gauge nylon liner has a soft nitrile foam palm for the ultimate comfort, dexterity, and sensitivity. Retail tag aids presentation for retail sales.
Colour: Grey/black
Certifications: EN ISO 21420: 2020 Dexterity 5, EN 388: 2016 + A1: 2018 4131X. ANSI/ISEA 105: 2016 CUT Level A1, ANSI/ISEA 105: 2016 ABRASION Level 4. CE certified.
Specifications
- Catalog No:
- PRTWA352G8RS
- PRTWA352G8RXXL
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Material:
- Nylon/polyester
- Nylon/polyester