Order Entry
Czech Republic
ContactUsLinkComponent
XELA UHPLC Columns, HILIC-Si
XELA UHPLC Columns, HILIC-Si
Supplier:  ISERA
Certificates
XELA UHPLC Columns, HILIC-Si
Supplier:  ISERA

Specifications

  • Catalog No:
  • ISERA211-K11X00
  • ISERA211-K11A00
  • ISERA211-K11Z00
  • ISERA211-K11U00
  • ISERA211-K11V00
  • ISERA211-K11M00
  • ISERA211-K11H00
  • ISERA211-K11K00
  • ISERA211-K11F00
  • ISERA211-K11G00
  • ISERA211-K11B00
  • ISERA211-K11D00
  • Phase name:
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • XELA
  • Max. pressure:
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • 1000 bar
  • pH range:
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • 2,5 - 7
  • USP code:
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • L3
  • Storage:
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • Shipping conditions:
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.

Specifications

Frequently Bought Together