Order Entry
Czech Republic
ContactUsLinkComponent
Metab-AAC HPLC Columns, BF Series
Metab-AAC HPLC Columns, BF Series
Supplier:  ISERA
Certificates
Metab-AAC HPLC Columns, BF Series
Supplier:  ISERA

Specifications

  • Catalog No:
  • ISERA1BF-A1AA0N
  • ISERA1BF-A1AB0N
  • ISERA1BF-A3AG0N
  • ISERA1BF-A2AG0N
  • ISERA1BF-A4BG0N
  • ISERA1BF-A1AG0N
  • ISERA1BF-A4BA0N
  • ISERA1BF-A3AA0N
  • ISERA1BF-A3AB0N
  • ISERA1BF-A2AA0N
  • ISERA1BF-A2AB0N
  • Description:
  • ISERA1BF-A1AA0N
  • ISERA1BF-A1AB0N
  • ISERA1BF-A3AG0N
  • ISERA1BF-A2AG0N
  • ISERA1BF-A4BG0N
  • ISERA1BF-A1AG0N
  • ISERA1BF-A4BA0N
  • ISERA1BF-A3AA0N
  • ISERA1BF-A3AB0N
  • ISERA1BF-A2AA0N
  • ISERA1BF-A2AB0N
  • Phase name:
  • Metab
  • Metab
  • Metab
  • Metab
  • Metab
  • Metab
  • Metab
  • Metab
  • Metab
  • Metab
  • Metab
  • Application:
  • Determination of organic acids, carbohydrates and alcohols in food and fermentation products
  • Determination of organic acids, carbohydrates and alcohols in food and fermentation products
  • Guard column
  • Guard column
  • Guard column
  • Guard column
  • Determination of oligosaccharides
  • Determination of mono- and disaccharides (and other)
  • Determination of mono- and disaccharides (and other)
  • Determination of sugar and sugar alcohols (and other)
  • Determination of sugar and sugar alcohols (and other)
  • Max. pressure:
  • 100 bar
  • 100 bar
  • 100 bar
  • 100 bar
  • 100 bar
  • 100 bar
  • 100 bar
  • 100 bar
  • 100 bar
  • 100 bar
  • pH range:
  • 0 - 14
  • 0 - 14
  • Neutral
  • 5 - 9
  • 5 - 9
  • 5 - 9
  • 5 - 9
  • USP code:
  • L17
  • L17
  • L17
  • L17
  • L17
  • L17
  • L17
  • L17
  • L17
  • L17
  • L17
  • Storage:
  • 4...25 °C
  • 5...25 °C
  • 12...25 °C
  • 11...25 °C
  • 13...25 °C
  • 14...25 °C
  • 10...25 °C
  • 8...25 °C
  • 9...25 °C
  • 6...25 °C
  • 7...25 °C
  • Shipping conditions:
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.

Specifications

Guard column holders

Product Information