Order Entry
Czech Republic
ContactUsLinkComponent
ISAspher HPLC Columns, C30-CXT
ISAspher HPLC Columns, C30-CXT
Supplier:  ISERA
Certificates
ISAspher HPLC Columns, C30-CXT
Supplier:  ISERA

Specifications

  • Catalog No:
  • ISERA111-L55A00
  • ISERA111-L55Y00
  • ISERA112-L55G30
  • ISERA111-L55B00
  • ISERA111-L55Z00
  • ISERA111-L55W00
  • ISERA112-L55E30
  • ISERA111-L55X00
  • ISERA112-L55F30
  • ISERA112-L55B30
  • ISERA112-L55C30
  • ISERA111-L55V00
  • ISERA111-L55T00
  • ISERA112-L55A30
  • ISERA111-L55U00
  • ISERA111-L55R00
  • ISERA111-L55S00
  • ISERA111-L55P00
  • ISERA111-L55Q00
  • ISERA111-L55M00
  • ISERA111-L55N00
  • ISERA111-L55L00
  • ISERA111-L55J00
  • ISERA111-L55K00
  • ISERA111-L55G00
  • ISERA111-L55H00
  • ISERA111-L55E00
  • ISERA111-L55F00
  • ISERA111-L55C00
  • ISERA111-L55D00
  • Phase name:
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • ISAspher
  • Application:
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Amongs other things, suitable for the separation of long-chain isomers (e.g. carotenoids, xanthophylls, tetraterpenes)
  • Max. pressure:
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • 400 bar
  • pH range:
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • 2,5 - 8
  • USP code:
  • L62
  • L62
  • L62
  • L62
  • L62
  • L62
  • L62
  • L62
  • L62
  • L62
  • L62
  • L62
  • L62
  • L62
  • L62
  • L62
  • L62
  • L62
  • L62
  • L62
  • L62
  • L62
  • L62
  • L62
  • L62
  • L62
  • L62
  • L62
  • L62
  • L62
  • Storage:
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • RT
  • Shipping conditions:
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.
  • Shipped under testing eluent.

Specifications

Guard column holders

Product Information

Frequently Bought Together