Matte Amber Ultra Heat Resistant Polyimide Labels for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Dodavatel: Brady
Certifikáty
O tomto produktu
Amber high temperature polyimide with an ultra-durable adhesive withstands the extreme wash protocol and cleaning chemicals used in printed circuit board manufacturing.
- Withstands multiple cycles of harsh condition washes
- Resists high temperatures up to 300 °C (572 °F)
- Survives on the top or bottom side of printed circuit boards through the solder process
- Matte finish
Ideal for circuit board labelling and component and equipment labelling.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R4310 for the M610/BMP61/BradyPrinter M611/M611 label printers or M7-R4300 for the M710/BMP71 label printers. This label is no longer compatible with TLS series printers.
Certifications: Meets the performance requirements of MIL -STD-202G, Notice 12, Method 215K. Meets the requirements of SAE AS81531 Marking of Electrical Insulating Material.
Frequently Bought Together