Harsh Environment Multi-Purpose Polyester Labels for M610, M611, BMP61, M710 (with Media Adapter) and BMP71 (with Media Adapter)
Dodavatel: Brady
Certifikáty
O tomto produktu
Permanent polyester (B-423) is ideal for electronic PCB and component identification, bar code labels, rating plates, and solar panel identification.
- Glossy white finish with permanent acrylic is thermal transfer printable
- Service temperature of −70 to 110 °C (−94 to 230 °F)
Ideal for barcode labelling, circuit board labelling, component and equipment labelling, general identification, and inventory and inspection labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R6010 for the M610/BMP61/BradyPrinter M611/M611 label printers or M7-R6000 for the M710/BMP71 label printers. This label is no longer compatible with TLS series printers.
Certifications: UL recognised. CSA accepted. Halogen-free (DIN VDE 0472/part 815). Approvals are for White material only.
Frequently Bought Together