Clear Welding Visors/Plates
Catalog # MMMAH523001
Supplier: 3M
Specifications
- Description:Clear welding visors plate, 9100, anti-fog
- Pk:5
Specifications
About this item
To ensure effective protection, replace pitted, scratched, or otherwise worn or damaged visors and plates.
- Replacement parts for original plates and visors
- Protect the welding lens from welding sparks and spatter
- Replace when parts become deeply pitted or too dirty to clean
- Regularly clean with a lint-free cloth
Get protection from welding sparks and spatter with 3M™ Welding clear visors and plates. Increase the service life of equipment by checking it regularly. Visor plates and shields should be replaced when scratched, pitted, or too dirty to clean. Regularly clean with a lint-free cloth, for example the 3M™ cleaning wipe.