Speedglas™ Inner Cover Plates
Catalog # MMMAH528015
Supplier: 3M
Specifications
- Description:Speedglas™ Inner cover plate, 9100X
- Seq:01
- Pk:5
Specifications
About this item
Speedglas™ Inner protection plates are replaceable spare parts for 3M™ Speedglas™ welding helmets and are designed to protect the welding lens.
- Helps protect the welding lens
- Replace inner cover plate when it becomes deeply pitted or too dirty to clean
- Regularly clean with a lint-free tissue or cloth
For maximum safety and comfort, regularly check equipment for worn or damaged parts.