ContactUsLinkComponent
Blood Bank Refrigerators, TSB
undefined
Blood Bank Refrigerators, TSB
  • Catalog No:
  • HERATSB140REGBV
  • HERATSB700REGBV
  • HERATSB250REGBV
  • HERATSB1500REGBV
  • HERATSB400REGBV
  • Temperature:
  • 4 °C (Setpoint)
  • 4 °C (Setpoint)
  • 4 °C (Setpoint)
  • 4 °C (Setpoint)
  • 4 °C (Setpoint)
  • Defrost:
  • Adaptive defrost
  • Adaptive defrost
  • Adaptive defrost
  • Adaptive defrost
  • Adaptive defrost
  • Max. load per shelf:
  • 30 kg
  • 30 kg
  • 30 kg
  • 30 kg
  • 30 kg
  • Refrigerant:
  • R290
  • R290
  • R290
  • R290
  • R290
  • Electrical:
  • 230 V/50 Hz
  • 230 V/50 Hz
  • 230 V/50 Hz
  • 230 V/50 Hz
  • 230 V/50 Hz
  • Plug:
  • EU
  • EU
  • EU
  • EU
  • EU
  • Shipping weight:
  • 87 kg
  • 215 kg
  • 131 kg
  • 283 kg
  • 150 kg
Frequently Bought Together