Folding magnifier, metal
Katalogové číslo 630-8231
Dodavatel: ESCHENBACH OPTIK
Specifikace
- Ø:21 mm
- Magnification:6×
- Colour:Silver/black
- Description:Folding metal magnifier with circular lens
- Pk:1
Specifikace
O tomto produktu
Metal folding magnifier with aplanatic lens and 6× magnification.
- Lens made of high quality silicate
- Matt chromium-plated brass body with eyelet
- Black painted metal lens frame
- Distortion-free illustration