Cole-Parmer® SHM-250 Digi-Mantle Heaters
Supplier: Cole-Parmer
Certificates
About this item
Heating mantles with interchangeable cartridges.
- Set speed and temperature in seconds with the intuitive display
- Complete system requires controller and heating cartridge
- Achieve uniform temperature up to 450 °C across round bottom flask
- Stirring speeds from 100 to 2000 min⁻¹ with excellent stability
- Vented casing keeps heating cartridge cool to the touch
- Modular design allows different size heating cartridges to be used on the same Digi-mantle controller
Control temperature and stirring speed independently on the Digi-Mantle digital controller. The control panel features indicator lights for power, heater and stirrer. Microprocessor controlled heater displays the heat setting as a percentage of the total power. View the actual temperature (up to 220 °C) of the flask content on a 3-digit LCD display by using the included temperature probe.
Delivery: Order controller and cartridges separately or heating kit includes a controller and a heating cartridge that accommodates 100 to 250, 500 or 1000 ml round bottom flask. Due to its modular design a different size heating cartridge can be used with the same controller, saving you money.
Specifications
- Catalog No:
- ELET36204-13
- ELET36204-09
- ELET36204-15
- ELET36204-05
- ELET36204-23
- ELET36204-19
- ELET36204-01
- Electrical:
- 230 VAC, 50 Hz
- 230 VAC, 50 Hz
- 230 VAC, 50 Hz
- 230 VAC, 50 Hz
- 230 VAC, 50 Hz
- 230 VAC, 50 Hz
- 230 VAC, 50 Hz
- Max. temperature:
- 450 °C
- 450 °C
- 450 °C
- 450 °C
- 450 °C
- 450 °C
- 450 °C