ContactUsLinkComponent
Reaction Stations, RS-200 and RS-250
undefined
Reaction Stations, RS-200 and RS-250
  • Catalog No:
  • ELET36630-01
  • ELET36630-11
  • Model:
  • RS-200
  • RS-250
  • Electrical:
  • 230 VAC, 50 Hz
  • 230 VAC, 50 Hz
  • Speed range:
  • 100 - 800 min⁻¹
  • 400 - 2000 min⁻¹
  • W×D×H:
  • 23,5×33×30,2 cm
  • 8×30,5×15 cm
  • Temperature range:
  • 220 °C
  • 150 °C
Frequently Bought Together