ContactUsLinkComponent
PUREZERO* HG5 SGX* Nitrile Gloves, Blue, Non Sterile
 :  Halyard
undefined
PUREZERO* HG5 SGX* Nitrile Gloves, Blue, Non Sterile
 :  Halyard
  • Catalog No:
  • 112-6502
  • 112-6503
  • 112-6504
  • 112-6505
  • 112-6506
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Cleanroom class:
  • 100/ISO 5
  • 100/ISO 5
  • 100/ISO 5
  • 100/ISO 5
  • 100/ISO 5
  • Material:
  • Nitrile
  • Nitrile
  • Nitrile
  • Nitrile
  • Nitrile
  • Finish:
  • Smooth SGX grip/textured fingertips
  • Smooth SGX grip/textured fingertips
  • Smooth SGX grip/textured fingertips
  • Smooth SGX grip/textured fingertips
  • Smooth SGX grip/textured fingertips
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Cuff thickness:
  • 0,07 mm (2,8 mil)
  • 0,07 mm (2,8 mil)
  • 0,07 mm (2,8 mil)
  • 0,07 mm (2,8 mil)
  • 0,07 mm (2,8 mil)
  • Finger thickness:
  • 0,10 mm (3,9 mil)
  • 0,10 mm (3,9 mil)
  • 0,10 mm (3,9 mil)
  • 0,10 mm (3,9 mil)
  • 0,10 mm (3,9 mil)
  • Palm thickness:
  • 0,08 mm (3,2 mil)
  • 0,08 mm (3,2 mil)
  • 0,08 mm (3,2 mil)
  • 0,08 mm (3,2 mil)
  • 0,08 mm (3,2 mil)
  • FDA regulated:
  • +
  • +
  • +
  • +
  • +
  • Tensile strength:
  • 24 MPa (Target)
  • 24 MPa (Target)
  • 24 MPa (Target)
  • 24 MPa (Target)
  • 24 MPa (Target)
  • EN norm:
  • EN ISO 374-1:2016+A1:2018/Type B Resistance to Chemical Permeation, EN ISO 374-5:2016 Micro-organism and Virus Protection, EN ISO 374-5:2016/ Protection against Micro-organism Risk, EN ISO 374-2:2019/ Resistance to Penetration, EN ISO 374-4:2019/ Resistance to Degradation, EN ISO 21420:2020/ General Requirement for Gloves
  • EN ISO 374-1:2016+A1:2018/Type B Resistance to Chemical Permeation, EN ISO 374-5:2016 Micro-organism and Virus Protection, EN ISO 374-5:2016/ Protection against Micro-organism Risk, EN ISO 374-2:2019/ Resistance to Penetration, EN ISO 374-4:2019/ Resistance to Degradation, EN ISO 21420:2020/ General Requirement for Gloves
  • EN ISO 374-1:2016+A1:2018/Type B Resistance to Chemical Permeation, EN ISO 374-5:2016 Micro-organism and Virus Protection, EN ISO 374-5:2016/ Protection against Micro-organism Risk, EN ISO 374-2:2019/ Resistance to Penetration, EN ISO 374-4:2019/ Resistance to Degradation, EN ISO 21420:2020/ General Requirement for Gloves
  • EN ISO 374-1:2016+A1:2018/Type B Resistance to Chemical Permeation, EN ISO 374-5:2016 Micro-organism and Virus Protection, EN ISO 374-5:2016/ Protection against Micro-organism Risk, EN ISO 374-2:2019/ Resistance to Penetration, EN ISO 374-4:2019/ Resistance to Degradation, EN ISO 21420:2020/ General Requirement for Gloves
  • EN ISO 374-1:2016+A1:2018/Type B Resistance to Chemical Permeation, EN ISO 374-5:2016 Micro-organism and Virus Protection, EN ISO 374-5:2016/ Protection against Micro-organism Risk, EN ISO 374-2:2019/ Resistance to Penetration, EN ISO 374-4:2019/ Resistance to Degradation, EN ISO 21420:2020/ General Requirement for Gloves
  • Sterility:
  • Non sterile
  • Non sterile
  • Non sterile
  • Non sterile
  • Non sterile
  • Anti-static:
  • +
  • +
  • +
  • +
  • +
  • Food contact approved:
  • +
  • +
  • +
  • +
  • +
Frequently Bought Together