ContactUsLinkComponent
Whatman™ 903 Proteinsaver Snap-Apart Sample Collection Card
Whatman™ 903 Proteinsaver Snap-Apart Sample Collection Card
Catalog # WHAT10534320
undefined
Whatman™ 903 Proteinsaver Snap-Apart Sample Collection Card
Catalog # WHAT10534320
Specifications
  • Description:
    Proteinsaver snap-apart cards
  • Pk:
    100
Specifications
About this item
903 sample collection card with four sample spots.
  • Ready-to-use cards and kits save time and costs during sample collection, transport, and archiving.
  • Strict quality control and ISO9001 manufacturing standards ensure high quality, reproducibility, and purity.
The paper in our 903 Proteinsaver Snap-Apart Card, imprinted with four half-inch circles, is enclosed between two pieces of cover stock. Each circle holds 75 - 80 µl of sample. To use the 903 Proteinsaver Snap-Apart Card, one cover is snapped off, the sample is collected and the remaining cover is folded over the sample. This cover is imprinted with the universal biohazard symbol.
The cards are ready to use and provide easy collection, identification, and transport of samples, without the need for costly freezers or dry ice delivery.
Certifications: Under current regulations, specimen collection cards, including neonatal screening cards, which are to be used for human diagnostic tests, are classified as 'other' IVD devices under the European IVD Directive and require the CE mark if sold within the European Union. All 903 specimen collection devices manufactured and printed by Whatman for neonatal testing in the EU undergo post-printing quality control and carry the CE mark.
Caution: Protein saver cards are for research use only. Whatman will assume no responsibility for the quality or performance of 903 collection devices converted, printed, or packaged by third party suppliers.