Order Entry
Czech Republic
ContactUsLinkComponent
E.Z.N.A.® Plasmid DNA Mini Kit I, (V-spin)
E.Z.N.A.® Plasmid DNA Mini Kit I, (V-spin)
Catalog # D6943-03
Supplier:  OMEGA BIO-TEK
CAS Number:  
E.Z.N.A.® Plasmid DNA Mini Kit I, (V-spin)
Catalog # D6943-03
Supplier:  OMEGA BIO-TEK
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Plasmid
  • Sample type:
    Bacteria
  • Target molecule:
    Plasmid
  • Description:
    Mini Plasmid Kit
  • Contents:
    600 Preps

Specifications

About this item

Isolate up to 25 µg plasmid DNA from 1 to 5 ml bacterial cultures using mini spin columns.

  • Rapid - Purification of plasmid DNA in less than 30 minutes
  • Safe - No Phenol/chloroform extractions
  • Versatile - Spin and vacuum formats available
  • High-quality - DNA is suitable for a variety of downstream applications

The E.Z.N.A.® Plasmid DNA Mini Kit I is designed to isolate up to 25 µg of high-quality plasmid DNA from 1 to 5 ml bacterial cultures in less than 30 minutes. Plasmid DNA purification follows the alkaline-lysis method and is simplified with HiBind® Mini Column technology into three quick steps: Bind, Wash, and Elute. Purified plasmid DNA is immediately ready for a wide variety of downstream applications such as routine screening, restriction enzyme digestion, transformation, PCR and DNA sequencing.