ContactUsLinkComponent
Tie-On Cleanroom Facemasks, Sterile, Tera 230
Tie-On Cleanroom Facemasks, Sterile, Tera 230
Catalog # 114-9594
Supplier:  ISOFIELD IRELAND
undefined
Tie-On Cleanroom Facemasks, Sterile, Tera 230
Catalog # 114-9594
Supplier:  ISOFIELD IRELAND
Specifications
  • Type:
    With ties
  • Colour:
    White
  • Size:
    Universal
  • Pk:
    500
Specifications
About this item
The Isofield Tera is the widest validated sterile facemask available.
  • ISO Class 4, GMP Grade A compatible
  • Four ties made from polyester
  • Validated sterile
  • No-smell gamma compatible materials
  • Excellent BFE and PFE
  • Malleable alluminium nose strip
Measuring 230 mm wide, with comfortable polyester ties, the Tera is made from gamma compatible materials ensuring low/no odour in use. The individually packed Tera facemasks provide excellent bacterial and particle filtration efficiency, and are GMP Grade A compatible.
Packaging: Each piece is sealed in HDPE bag. 50 sealed individual bags per inner bag, 10 inner bags per case (500 pieces).