ContactUsLinkComponent
Tie-On Cleanroom Facemasks, Non Sterile, Zetta 230
Tie-On Cleanroom Facemasks, Non Sterile, Zetta 230
Catalog # 114-9596
Supplier:  ISOFIELD IRELAND
undefined
Tie-On Cleanroom Facemasks, Non Sterile, Zetta 230
Catalog # 114-9596
Supplier:  ISOFIELD IRELAND
Specifications
  • Type:
    With ties
  • Colour:
    White
  • Size:
    Universal
  • Pk:
    500
Specifications
About this item
The Isofield Zetta facemask is ISO Class 4 compatible.
  • ISO Class 4 compatible
  • Extra wide at 230 mm
  • Four ties made from polyester
  • Individually packed
  • Low CFU
  • Excellent BFE and PFE
  • Malleable alluminium nose strip
Measuring 230 mm wide, with comfortable polyester ties, the Zetta is made from low-linting cleanroom grade material to ensure best in class cleanliness. The individually packed Zetta facemasks provide excellent bacterial and particle filtration efficiency.
Packaging: Each piece is sealed in HDPE bag. 50 sealed individual bags per inner bag, 10 inner bags per case (500 pieces).