Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Microbiology Test Systems
Microorganism Tests
Culti-Loops™
Staphylococcus aureus
ATCC™ 25923™
Culti-Loops™ Staphylococcus aureus ATCC™ 25923™
Culti-Loops™ Staphylococcus aureus ATCC™ 25923™
Catalog # OXOIR4607010
Supplier:
THERMO FISHER DIAGNOSTICS
CAS Number:
Expand image
Culti-Loops™ Staphylococcus aureus ATCC™ 25923™
Catalog # OXOIR4607010
Supplier:
THERMO FISHER DIAGNOSTICS
CAS Number:
Specifications
Pk:
5
Description:
Staphylococcus aureus ATCC™ 25923™
Specifications