Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Baths
Tissue Baths
Accessories for Tissue Float Bath, Cole-Parmer®, FB-200
Accessories for Tissue Float Bath, Cole-Parmer®, FB-200
Accessories for Tissue Float Bath, Cole-Parmer®, FB-200
Katalogové číslo ELET48572-99
Supplier:
Cole-Parmer
Expand image
Accessories for Tissue Float Bath, Cole-Parmer®, FB-200
Katalogové číslo ELET48572-99
Supplier:
Cole-Parmer
Specifications
Description:
Lid for tissue float bath
Pk:
1
Specifications
About this item
Product Details
Product Details