ContactUsLinkComponent
Bole white (kaolin), powder FCC
Bole white (kaolin), powder FCC
  • Acid-Soluble Substances:
    max. 2.0%
  • Arsenic (As):
    max.3 mg/kg
  • Carbonate (CO₃):
    To Pass Test
  • Iron (Fe):
    To Pass Test
  • Lead (Pb):
    max. 10 mg/kg
Frequently Bought Together