D-(+)-Glucosamine hydrochloride ≥98%
Supplier: Spectrum Chemical
Certificates
About this item
Synonyms: 2-Amino-2-deoxy-D-glucopyranose Hydrochloride, D-Glucosamine hydrochloride, (2R,3R,4S,5R)-2-Amino-3,4,5,6-tetrahydroxyhexanal hydrochloride
CAS Number:66-84-2
:
Formula:C₆H₁₃NO₅
MDL Number:MFCD00135831
Molecular Weight:215.63 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:98% min.
- Moisture:max.1.5 %
- Heavy Metals:max. 10 ppm
Specification Test Results
Frequently Bought Together