D-Lactose monohydrate, powder NF
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:64044-51-5
:63-42-3
Formula:C₁₂H₂₂O₁₁·1H₂O
MDL Number:MFCD00150747
:360.32 g/mol
Storage Temperature:Ambient
Specification Test Results
- Clarity and Color of Solution:To Pass Test
- Specific Rotation [A]D:+54.4 to +55.9
- Acidity / Alkalinity:To Pass Test
- Water:4.5 - 5.5 %
- Loss On Drying:max. 0.5 %
Specification Test Results
Frequently Bought Together