Order Entry
Czech Republic
ContactUsLinkComponent
4,4'-Dimethoxytrityl chloride ≥98%
4,4'-Dimethoxytrityl chloride ≥98%
Catalog # SPCMD2181-5KGBL
Supplier:  Spectrum Chemical
CAS Number:  
4,4'-Dimethoxytrityl chloride ≥98%
Catalog # SPCMD2181-5KGBL
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pk:
    5 kg
  • Seq:
    4114

Specifications

Specification Test Results

  • Assay:
    Approx. 98%

Specification Test Results

About this item

 :
 :338.8 g/mol
Storage Temperature:Ambient
CAS Number:40615-36-9
Formula:C₂₁H₁₉ClO₂
MDL Number:MFCD00008409