8-Anilino-1-naphthalenesulphonic acid ≥95%
Supplier: Spectrum Chemical
Certificates
About this item
Synonyms: 8-Anilinonaphthalene-1-sulphonic acid, 8-(Phenylamino)naphthalene-1-sulphonic acid
CAS Number:82-76-8
:
Formula:C₁₆H₁₃NO₃S
MDL Number:MFCD00003998
Molecular Weight:299.35 g/mol
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:95% min.
Specification Test Results
Frequently Bought Together