Dextrin, white powder
Supplier: Spectrum Chemical
Certificates
About this item
Synonyms: Dextrin
CAS Number:9004-53-9
:
Formula:(C₆H₁₂O₆)n
MDL Number:MFCD00081554
Molecular Weight:180.16 g/mol
Merck Index:13,02971
Storage Temperature:Ambient
EINECS:232-675-4
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together