Propyl-3,4,5-trihydroxybenzoate ≥98% NF
Supplier: Spectrum Chemical
Certificates
About this item
Synonyms: Gallic acid propyl ester, Propyl gallate, Propyl 3,4,5-trihydroxybenzoate, Progalline P, N-Propyl gallate
CAS Number:121-79-9
:
Formula:C₁₀H₁₂O₅
MDL Number:MFCD00002196
:212.2 g/mol
Merck Index:12,08044
Boiling Pt:449 °C (1013 hPa)
Melting Pt:145…148 °C
Density:1,21 g/cm³ (20 °C)
EINECS:204-498-2
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:98% Min.
Specification Test Results
Frequently Bought Together