


Diethylenetriamine ≥98%
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:111-40-0
:
Formula:C₄H₁₃N₃
MDL Number:MFCD00008171
:103.17 g/mol
Density:0,955
UN:2079
ADR:8,II
:GHS05
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Assay:98% min.
- Water:max. 0.5 %
Specification Test Results
Frequently Bought Together